Product Details

SNP ID
rs10843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31528709 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCCTAGCTCAGGGACCCAGCCCC[T/C]CCTCTCTGAGAAACTCTGACCTTCA
Phenotype
MIM: 605821
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
AHSP PubMed Links

Gene Details

Gene
AHSP
Gene Name
alpha hemoglobin stabilizing protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318221.1 442 UTR 3 NP_001305150.1
NM_001318222.1 442 UTR 3 NP_001305151.1
NM_016633.3 442 UTR 3 NP_057717.1

View Full Product Details