Product Details

SNP ID
rs212433
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:105879837 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCACATTTAATTGATAACAGAGTAT[A/G]TCTACATTGTTGCAGTTAGTTATGG
Phenotype
MIM: 615610
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATXN7L1 PubMed Links

Gene Details

Gene
ATXN7L1
Gene Name
ataxin 7 like 1
There are no transcripts associated with this gene.

Gene
CDHR3
Gene Name
cadherin related family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301161.1 Intron NP_001288090.1
NM_152750.4 Intron NP_689963.2
XM_005250224.4 Intron XP_005250281.1
XM_005250225.2 Intron XP_005250282.1
XM_006715905.2 Intron XP_006715968.1
XM_011515955.2 Intron XP_011514257.1
XM_011515956.2 Intron XP_011514258.1
XM_011515957.2 Intron XP_011514259.1
XM_011515958.2 Intron XP_011514260.1
XM_011515959.2 Intron XP_011514261.1
XM_017011862.1 Intron XP_016867351.1
XM_017011863.1 Intron XP_016867352.1
XM_017011864.1 Intron XP_016867353.1
XM_017011865.1 Intron XP_016867354.1
XM_017011866.1 Intron XP_016867355.1
XM_017011867.1 Intron XP_016867356.1

View Full Product Details