Product Details

SNP ID
rs4257849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:37443193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTATATCGTTCCTTCCAAAAGC[G/T]CTTTTTGCAATTATAAGAAATAAAA
Phenotype
MIM: 616189
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CMTR1 PubMed Links

Gene Details

Gene
CMTR1
Gene Name
cap methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015050.2 Intron NP_055865.1
XM_005248955.3 Intron XP_005249012.1
Gene
LOC107986531
Gene Name
oleosin-B6-like
There are no transcripts associated with this gene.

View Full Product Details