Product Details

SNP ID
rs13219624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:34894022 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTATCTAGAAAGCACAGTTATCCA[A/G]AATGTCTAGGCTTACAGAGGCTCTG
Phenotype
MIM: 608994 MIM: 600772
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKS1A PubMed Links

Gene Details

Gene
ANKS1A
Gene Name
ankyrin repeat and sterile alpha motif domain containing 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015245.2 Intron NP_056060.2
XM_005248964.3 Intron XP_005249021.1
XM_006715036.2 Intron XP_006715099.1
XM_011514431.2 Intron XP_011512733.1
XM_011514432.2 Intron XP_011512734.1
XM_011514433.1 Intron XP_011512735.1
XM_011514434.2 Intron XP_011512736.1
XM_011514435.2 Intron XP_011512737.1
XM_011514436.2 Intron XP_011512738.1
XM_011514437.2 Intron XP_011512739.1
XM_017010593.1 Intron XP_016866082.1
Gene
LOC105375030
Gene Name
uncharacterized LOC105375030
There are no transcripts associated with this gene.

Gene
TAF11
Gene Name
TATA-box binding protein associated factor 11
There are no transcripts associated with this gene.

View Full Product Details