Product Details

SNP ID
rs10407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50185622 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGGTGGTTTTGTATTCAATCAC[A/T]GTCTTGCCCCAGGCTCCGGTGTGAC
Phenotype
MIM: 120150 MIM: 600119
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
COL1A1 PubMed Links

Gene Details

Gene
COL1A1
Gene Name
collagen type I alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000088.3 4212 Silent Mutation ACA,ACT T1425T NP_000079.2
XM_005257058.3 4212 Silent Mutation ACA,ACT T1335T XP_005257115.2
XM_005257059.4 4212 Silent Mutation ACA,ACT T1119T XP_005257116.2
XM_011524341.1 4212 Silent Mutation ACA,ACT T1359T XP_011522643.1
Gene
SGCA
Gene Name
sarcoglycan alpha
There are no transcripts associated with this gene.

View Full Product Details