Product Details

SNP ID
rs1878388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42609610 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGTGCCATTCATTGCCAGAACTCG[A/G]TTCTCATTGGAGCGGAGGAAGTTCG
Phenotype
MIM: 616498 MIM: 191135
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM134C PubMed Links

Gene Details

Gene
FAM134C
Gene Name
family with sequence similarity 134 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178126.3 271 Intron NP_835227.1
XM_011524440.2 271 Intron XP_011522742.1
Gene
TUBG1
Gene Name
tubulin gamma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001070.4 271 UTR 5 NP_001061.2

View Full Product Details