Product Details
- SNP ID
-
rs1997960
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:152971577 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTAACTATTGCCCTTTATCTATGCA[G/T]TATCTGATAGATGCTCAATAAATGC
- Phenotype
-
MIM: 606013
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FBXO5
PubMed Links
Gene Details
- Gene
- FBXO5
- Gene Name
- F-box protein 5
View Full Product Details