Product Details

SNP ID
rs2030671
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:70250837 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTCTGCATAAATGGGGGTAAGCTCT[A/G]AAGGGGAGAAGAGGTGATTAAAGCA
Phenotype
MIM: 604909
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNOT2 PubMed Links

Gene Details

Gene
CNOT2
Gene Name
CCR4-NOT transcription complex subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199302.1 Intron NP_001186231.1
NM_001199303.1 Intron NP_001186232.1
NM_014515.5 Intron NP_055330.1
XM_006719429.3 Intron XP_006719492.1
XM_006719430.3 Intron XP_006719493.1
XM_006719432.3 Intron XP_006719495.1
XM_006719433.3 Intron XP_006719496.1
XM_011538399.1 Intron XP_011536701.1
XM_011538400.2 Intron XP_011536702.1
XM_017019348.1 Intron XP_016874837.1
XM_017019349.1 Intron XP_016874838.1
XM_017019350.1 Intron XP_016874839.1
XM_017019351.1 Intron XP_016874840.1
XM_017019352.1 Intron XP_016874841.1
XM_017019353.1 Intron XP_016874842.1
XM_017019354.1 Intron XP_016874843.1
Gene
LINC01481
Gene Name
long intergenic non-protein coding RNA 1481
There are no transcripts associated with this gene.

View Full Product Details