Product Details

SNP ID
rs1877686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74274443 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTAATAACCACCAAGAGTGCCAAA[A/C]ATTCCTCTTGGGGGGCTTCCTTCTG
Phenotype
MIM: 605483
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DNAI2 PubMed Links

Gene Details

Gene
DNAI2
Gene Name
dynein axonemal intermediate chain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172810.1 Intron NP_001166281.1
NM_023036.4 Intron NP_075462.3
XM_011525125.2 Intron XP_011523427.1

View Full Product Details