Product Details

SNP ID
rs1985961
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:64039902 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGTCACGTTCTTTGTGGCTTTCCAA[T/C]GTCTACAGAAAGAATTTAAGCGCCG
Phenotype
MIM: 604033
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
ERN1 PubMed Links

Gene Details

Gene
ERN1
Gene Name
endoplasmic reticulum to nucleus signaling 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001433.3 8487 Intron NP_001424.3
XM_017024347.1 8487 UTR 3 XP_016879836.1
XM_017024348.1 8487 Intron XP_016879837.1

View Full Product Details