Product Details

SNP ID
rs1289389
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.13:98684036 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTCCTGCCTCCTTTAGAATAGTTTT[C/T]GTAAGAGTATTAGAAATGAACAGTA
Phenotype
MIM: 600544
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC15A1 PubMed Links

Gene Details

Gene
SLC15A1
Gene Name
solute carrier family 15 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005073.3 2892 UTR 3 NP_005064.1

View Full Product Details