Product Details

SNP ID
rs1964243
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:68239165 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGGAGAAAACAGGTCACCTGCTGCT[G/T]CTTTGCATAGAGCCCTGGCTCTGAG
Phenotype
MIM: 615169
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AMZ2 PubMed Links

Gene Details

Gene
AMZ2
Gene Name
archaelysin family metallopeptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033569.1 Intron NP_001028741.1
NM_001033570.1 Intron NP_001028742.1
NM_001033571.1 Intron NP_001028743.1
NM_001033572.1 Intron NP_001028744.1
NM_001033574.1 Intron NP_001028746.1
NM_001289054.1 Intron NP_001275983.1
NM_001289056.1 Intron NP_001275985.1
NM_016627.4 Intron NP_057711.3
XM_005257432.4 Intron XP_005257489.1
XM_005257433.3 Intron XP_005257490.1
XM_005257435.4 Intron XP_005257492.1
XM_005257436.3 Intron XP_005257493.1
XM_006721940.2 Intron XP_006722003.1
XM_017024723.1 Intron XP_016880212.1
XM_017024724.1 Intron XP_016880213.1
XM_017024725.1 Intron XP_016880214.1
XM_017024726.1 Intron XP_016880215.1
XM_017024727.1 Intron XP_016880216.1
XM_017024728.1 Intron XP_016880217.1

View Full Product Details