Product Details
- SNP ID
-
hCV11432519
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:24812589 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACATCTGAAAAGAAAAAACAAAAA[C/T]CACTCTAAGTGTCCAAATATTGGAA
- Phenotype
-
MIM: 607958
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
STXBP6
PubMed Links
Gene Details
- Gene
- STXBP6
- Gene Name
- syntaxin binding protein 6
There are no transcripts associated with this gene.
View Full Product Details