Product Details

SNP ID
rs13487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20801418 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGGGGTTAACATGGGGCCCACC[A/T]CACAACCCACTCTTCACCCCCAAAA
Phenotype
MIM: 180440
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RNASE1 PubMed Links

Gene Details

Gene
RNASE1
Gene Name
ribonuclease A family member 1, pancreatic
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002933.4 828 UTR 3 NP_002924.1
NM_198232.2 828 UTR 3 NP_937875.1
NM_198234.2 828 UTR 3 NP_937877.1
NM_198235.2 828 UTR 3 NP_937878.1

View Full Product Details