Product Details
- SNP ID
-
rs2024857
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:26877048 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTTCTCTCTGTCCCTAACTCTCT[A/C]CTTTTCCCCTCCATAAATACATTTT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4307HG
PubMed Links
Gene Details
- Gene
- MIR4307HG
- Gene Name
- MIR4307 host gene
There are no transcripts associated with this gene.
View Full Product Details