Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001256732.1 | 1268 | UTR 3 | NP_001243661.1 | ||
NM_001256733.1 | 1268 | UTR 3 | NP_001243662.1 | ||
NM_001256734.1 | 1268 | UTR 3 | NP_001243663.1 | ||
NM_001256735.1 | 1268 | UTR 3 | NP_001243664.1 | ||
NM_001256736.1 | 1268 | UTR 3 | NP_001243665.1 | ||
NM_012446.3 | 1268 | UTR 3 | NP_036578.2 | ||
XM_011543297.2 | 1268 | UTR 3 | XP_011541599.1 | ||
XM_017009304.1 | 1268 | UTR 3 | XP_016864793.1 | ||
XM_017009305.1 | 1268 | UTR 3 | XP_016864794.1 | ||
XM_017009306.1 | 1268 | UTR 3 | XP_016864795.1 | ||
XM_017009307.1 | 1268 | UTR 3 | XP_016864796.1 | ||
XM_017009308.1 | 1268 | UTR 3 | XP_016864797.1 | ||
XM_017009309.1 | 1268 | UTR 3 | XP_016864798.1 | ||
XM_017009310.1 | 1268 | UTR 3 | XP_016864799.1 |