Product Details

SNP ID
rs1867661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:79244610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGACAGTAGGTCTTGAAAGCAC[A/C]AGATGAGCCCTCTGTTGCTCCATTG
Phenotype
MIM: 604279
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
JMY PubMed Links

Gene Details

Gene
JMY
Gene Name
junction mediating and regulatory protein, p53 cofactor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152405.4 Intron NP_689618.4
XM_005248430.2 Intron XP_005248487.1
XM_011543155.2 Intron XP_011541457.1

View Full Product Details