Product Details

SNP ID
rs12666309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:123772910 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGCAGATGTTACAGTTAAAAAACG[A/G]AATCTGATGTCAGACCAGACTACAC
Phenotype
MIM: 604510
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HYAL4 PubMed Links

Gene Details

Gene
HYAL4
Gene Name
hyaluronoglucosaminidase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012269.2 Intron NP_036401.2
XM_011515990.2 Intron XP_011514292.1
XM_011515991.2 Intron XP_011514293.1
XM_017011911.1 Intron XP_016867400.1

View Full Product Details