Product Details

SNP ID
rs13773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23568463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCACCCACCTGTCACCCGGGTTTG[A/G]CTCCCACCTCTGCCCTGCCTGGGAA
Phenotype
MIM: 603534 MIM: 611612
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP1G2 PubMed Links

Gene Details

Gene
AP1G2
Gene Name
adaptor related protein complex 1 gamma 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282474.1 594 Intron NP_001269403.1
NM_001282475.1 594 Intron NP_001269404.1
NM_003917.4 594 Intron NP_003908.1
XM_005268167.3 594 Intron XP_005268224.1
XM_005268168.4 594 Intron XP_005268225.1
XM_005268169.3 594 Intron XP_005268226.1
XM_005268170.3 594 Intron XP_005268227.1
XM_005268172.3 594 Intron XP_005268229.1
XM_005268173.3 594 Intron XP_005268230.1
XM_005268174.3 594 Intron XP_005268231.1
XM_005268175.3 594 Intron XP_005268232.1
XM_005268177.3 594 Intron XP_005268234.1
XM_005268178.3 594 Intron XP_005268235.1
XM_005268179.3 594 Intron XP_005268236.1
XM_005268180.4 594 Intron XP_005268237.1
XM_005268182.3 594 Intron XP_005268239.1
XM_006720301.3 594 Intron XP_006720364.1
XM_011537283.2 594 Intron XP_011535585.1
XM_011537286.2 594 Intron XP_011535588.1
XM_017021740.1 594 Intron XP_016877229.1
XM_017021741.1 594 Intron XP_016877230.1
XM_017021742.1 594 Intron XP_016877231.1
XM_017021743.1 594 Intron XP_016877232.1
XM_017021744.1 594 Intron XP_016877233.1
XM_017021745.1 594 Intron XP_016877234.1
XM_017021746.1 594 Intron XP_016877235.1
XM_017021747.1 594 UTR 5 XP_016877236.1
XM_017021748.1 594 Intron XP_016877237.1
Gene
JPH4
Gene Name
junctophilin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146028.1 594 UTR 3 NP_001139500.1
NM_032452.2 594 UTR 3 NP_115828.2
Gene
LOC102724814
Gene Name
uncharacterized LOC102724814
There are no transcripts associated with this gene.

Gene
THTPA
Gene Name
thiamine triphosphatase
There are no transcripts associated with this gene.

View Full Product Details