Product Details

SNP ID
rs2052579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:72010106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGGATCCTTAAAAGCAAATCT[A/G]ATCATGTCATTTCCTGGCTTAGGGA
Phenotype
MIM: 126064 MIM: 607895
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHODH PubMed Links

Gene Details

Gene
DHODH
Gene Name
dihydroorotate dehydrogenase (quinone)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001361.4 Intron NP_001352.2
XM_005255827.3 Intron XP_005255884.1
XM_005255829.3 Intron XP_005255886.1
XM_017022990.1 Intron XP_016878479.1
Gene
PKD1L3
Gene Name
polycystin 1 like 3, transient receptor potential channel interacting
There are no transcripts associated with this gene.

View Full Product Details