Product Details

SNP ID
rs12249
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:1232955 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACGTGCTCTCGGCCGACCTGGTGC[G/A]CTACCTGCGCCTCAGCCGCGACTAC
Phenotype
MIM: 615291 MIM: 616593 MIM: 614282
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
B3GALT6 PubMed Links

Gene Details

Gene
B3GALT6
Gene Name
beta-1,3-galactosyltransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080605.3 707 Missense Mutation CAC,CGC H226R NP_542172.2
Gene
FAM132A
Gene Name
family with sequence similarity 132 member A
There are no transcripts associated with this gene.

Gene
SDF4
Gene Name
stromal cell derived factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016176.3 707 Intron NP_057260.2
NM_016547.2 707 Intron NP_057631.1
XM_011541556.1 707 Intron XP_011539858.1

View Full Product Details