Product Details

SNP ID
rs1936044
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:112395427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCATTCCTGTGTTCACAAGGCA[C/G]ACATTCCTTGCATCCTCTGTACTCT
Phenotype
MIM: 615100
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CTTNBP2NL PubMed Links

Gene Details

Gene
CTTNBP2NL
Gene Name
CTTNBP2 N-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018704.2 Intron NP_061174.1

View Full Product Details