Product Details

SNP ID
hCV11451611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:82811015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATGCCTTCTAAGTCCACACCCAC[G/A]GTTTATTAAGGAGAGCCAGGCTGGA
Phenotype
MIM: 612393
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
FSD2 PubMed Links

Gene Details

Gene
FSD2
Gene Name
fibronectin type III and SPRY domain containing 2
There are no transcripts associated with this gene.

Gene
WHAMM
Gene Name
WAS protein homolog associated with actin, golgi membranes and microtubules
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080435.2 Intron NP_001073904.1

View Full Product Details