Product Details

SNP ID
rs1055986
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:17315757 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGAGGGTGGGATGGACAGGGAGGA[A/G]GAAGAGGGAGGGATTACAAAGTAGC
Phenotype
MIM: 610139
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ST8SIA6 PubMed Links

Gene Details

Gene
ST8SIA6
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004470.1 6310 Intron NP_001004470.1
XM_017016174.1 6310 UTR 3 XP_016871663.1
XM_017016175.1 6310 Intron XP_016871664.1

View Full Product Details