Product Details

SNP ID
rs1916341
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:73911404 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CAGGCTGCCCGGAGTCCGGAGCCCA[G/T]AGAGGAGAGAGACAGCTGGGGAGCC
Phenotype
MIM: 191840
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C10orf55 PubMed Links

Gene Details

Gene
C10orf55
Gene Name
chromosome 10 open reading frame 55
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001791.2 1078 UTR 3 NP_001001791.2
Gene
PLAU
Gene Name
plasminogen activator, urokinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145031.2 1078 Intron NP_001138503.1
NM_001319191.1 1078 Intron NP_001306120.1
NM_002658.4 1078 Intron NP_002649.1
XM_011539866.2 1078 Intron XP_011538168.1

View Full Product Details