Product Details

SNP ID
rs1042600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:200855038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAAACCCAAGATAGTATTCAATA[A/T]GAATGCATCCTATGCTCCAGACATC
Phenotype
MIM: 601951
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CLK1 PubMed Links

Gene Details

Gene
CLK1
Gene Name
CDC like kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162407.1 1287 Missense Mutation CAT,CTT H411L NP_001155879.1
NM_004071.3 1287 Missense Mutation CAT,CTT H369L NP_004062.2

View Full Product Details