Product Details

SNP ID
rs836899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:20188035 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCAGGGCTTTGCTCCGTAGTTAA[A/G]ACACTAGGGTATGTTTTTGTAGTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105372310 PubMed Links

Gene Details

Gene
LOC105372310
Gene Name
uncharacterized LOC105372310
There are no transcripts associated with this gene.

Gene
ZNF486
Gene Name
zinc finger protein 486
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052852.3 Intron NP_443084.2

View Full Product Details