Product Details

SNP ID
rs1989868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19336768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGTTCCTAATTTCAATTGTAGC[C/T]GAAATCTGCCTTCCTCCCACCTCAG
Phenotype
MIM: 614560
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAU2 PubMed Links

Gene Details

Gene
MAU2
Gene Name
MAU2 sister chromatid cohesion factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015329.3 Intron NP_056144.3
XM_005259837.3 Intron XP_005259894.2
XM_005259838.3 Intron XP_005259895.3
XM_006722711.3 Intron XP_006722774.2
XM_011527844.2 Intron XP_011526146.1
XM_017026539.1 Intron XP_016882028.1

View Full Product Details