Product Details

SNP ID
rs1865077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49636835 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAACCCCTGTCACCTCTGCCGGTC[A/G]TTAATGGCGAACACCAGCAGGAAGC
Phenotype
MIM: 165090
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RRAS PubMed Links

Gene Details

Gene
RRAS
Gene Name
related RAS viral (r-ras) oncogene homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006270.4 378 Silent Mutation AAC,AAT N111N NP_006261.1
Gene
SCAF1
Gene Name
SR-related CTD associated factor 1
There are no transcripts associated with this gene.

View Full Product Details