Product Details

SNP ID
rs1889785
Assay Type
Functionally tested
NCBI dbSNP Submissions
59
Location
Chr.1:16022234 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TGAGCTGTCGGGGAGAAAGGAGGGT[A/G]CTCCGAGATTGGCAGGGAGAGTCGG
Phenotype
MIM: 602024 MIM: 610692
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLCNKA PubMed Links

Gene Details

Gene
CLCNKA
Gene Name
chloride voltage-gated channel Ka
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042704.1 Intron NP_001036169.1
NM_001257139.1 Intron NP_001244068.1
NM_004070.3 Intron NP_004061.3
Gene
HSPB7
Gene Name
heat shock protein family B (small) member 7
There are no transcripts associated with this gene.

View Full Product Details