Product Details

SNP ID
rs6659086
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:25800934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGGACCGGGAGGCACTGGGAGA[A/G]CTCAGGAAGATGGTGGGAGAAGGGG
Phenotype
MIM: 606210
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEPN1 PubMed Links

Gene Details

Gene
SEPN1
Gene Name
selenoprotein N, 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020451.2 Intron NP_065184.2
NM_206926.1 Intron NP_996809.1

View Full Product Details