Product Details

SNP ID
rs590826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40000039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTTAGAAAAACTAATTCTCCCTA[A/G]CCATAGCTTTCTTATAACTCTGTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF546 PubMed Links

Gene Details

Gene
ZNF546
Gene Name
zinc finger protein 546
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297763.1 Intron NP_001284692.1
NM_178544.4 Intron NP_848639.2
XM_011526898.2 Intron XP_011525200.1
XM_011526899.2 Intron XP_011525201.1

View Full Product Details