Product Details

SNP ID
rs11621910
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94484511 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAAACTGCACACCTGCTGAAGCCC[A/G]CCAGAGTTCTTCAATTAAAGACCAG
Phenotype
MIM: 615677
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SERPINA12 PubMed Links

Gene Details

Gene
SERPINA12
Gene Name
serpin family A member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304461.1 Intron NP_001291390.1
NM_173850.3 Intron NP_776249.1
XM_011536451.2 Intron XP_011534753.1
XM_011536452.2 Intron XP_011534754.1
XM_011536453.2 Intron XP_011534755.1
XM_011536454.2 Intron XP_011534756.1
XM_011536455.1 Intron XP_011534757.1
XM_017020989.1 Intron XP_016876478.1
XM_017020990.1 Intron XP_016876479.1
Gene
SERPINA9
Gene Name
serpin family A member 9
There are no transcripts associated with this gene.

View Full Product Details