Product Details

SNP ID
rs2665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:71549302 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTCTTTTTCCAGCCCCTGAGGAAA[A/T]GGAATGAACCACTCCCTGCCCATTC
Phenotype
MIM: 148021
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP5-9 PubMed Links

Gene Details

Gene
KRTAP5-9
Gene Name
keratin associated protein 5-9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005553.3 883 UTR 3 NP_005544.4

View Full Product Details