Product Details

SNP ID
rs666092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73647340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAGCATCAAAAAAACATGCAAA[A/C]CTAGGAGCTTGGCTTCTCCTTGCAA
Phenotype
MIM: 607651
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PLEKHB1 PubMed Links

Gene Details

Gene
PLEKHB1
Gene Name
pleckstrin homology domain containing B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130033.1 85 Intron NP_001123505.1
NM_001130034.1 85 Intron NP_001123506.1
NM_001130035.1 85 Intron NP_001123507.1
NM_001130036.1 85 Intron NP_001123508.1
NM_021200.2 85 Intron NP_067023.1
XM_011545192.2 85 UTR 5 XP_011543494.1
XM_011545193.2 85 UTR 5 XP_011543495.1

View Full Product Details