Product Details

SNP ID
rs2052625
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:231041140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTTAAAGTTCGGTTTTAAAAAC[A/C]CACCCACTTTTGCAAATCAGACTCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C2orf72 PubMed Links

Gene Details

Gene
C2orf72
Gene Name
chromosome 2 open reading frame 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144994.1 Intron NP_001138466.1

View Full Product Details