Product Details

SNP ID
rs1062441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49184836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACTTTATTCAAAACCCATCACA[A/G]AAATGGACAGCTTGGGTCTGTAACA
Phenotype
MIM: 602529
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TUBA1A PubMed Links

Gene Details

Gene
TUBA1A
Gene Name
tubulin alpha 1a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270399.1 1963 UTR 3 NP_001257328.1
NM_001270400.1 1963 UTR 3 NP_001257329.1
NM_006009.3 1963 UTR 3 NP_006000.2

View Full Product Details