Product Details

SNP ID
rs14870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:35738780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAAAACAGTGCTTCATTACCATG[C/T]GCAAAGGCTGAGGCAGTGCTCCTCC
Phenotype
MIM: 607255 MIM: 612149
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APOL5 PubMed Links

Gene Details

Gene
APOL5
Gene Name
apolipoprotein L5
There are no transcripts associated with this gene.

Gene
RBFOX2
Gene Name
RNA binding protein, fox-1 homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031695.2 6957 UTR 3 NP_001026865.1
NM_001082576.1 6957 UTR 3 NP_001076045.1
NM_001082577.1 6957 UTR 3 NP_001076046.1
NM_001082578.1 6957 UTR 3 NP_001076047.1
NM_001082579.1 6957 UTR 3 NP_001076048.1
NM_014309.2 6957 UTR 3 NP_055124.1
XM_005261428.2 6957 UTR 3 XP_005261485.1
XM_005261429.2 6957 UTR 3 XP_005261486.1
XM_005261430.2 6957 UTR 3 XP_005261487.1
XM_005261431.2 6957 UTR 3 XP_005261488.1
XM_005261432.2 6957 UTR 3 XP_005261489.1
XM_005261433.2 6957 UTR 3 XP_005261490.1
XM_005261435.1 6957 UTR 3 XP_005261492.1
XM_005261437.1 6957 UTR 3 XP_005261494.1
XM_006724185.2 6957 UTR 3 XP_006724248.1
XM_006724186.2 6957 UTR 3 XP_006724249.1
XM_006724187.2 6957 UTR 3 XP_006724250.1
XM_006724188.2 6957 UTR 3 XP_006724251.1
XM_006724189.3 6957 UTR 3 XP_006724252.1
XM_006724190.1 6957 UTR 3 XP_006724253.1
XM_006724191.1 6957 UTR 3 XP_006724254.1
XM_006724192.2 6957 UTR 3 XP_006724255.1
XM_006724193.3 6957 Intron XP_006724256.1
XM_006724194.2 6957 UTR 3 XP_006724257.1
XM_017028686.1 6957 UTR 3 XP_016884175.1
XM_017028687.1 6957 UTR 3 XP_016884176.1
XM_017028688.1 6957 UTR 3 XP_016884177.1
XM_017028689.1 6957 UTR 3 XP_016884178.1
XM_017028690.1 6957 UTR 3 XP_016884179.1
XM_017028691.1 6957 UTR 3 XP_016884180.1
XM_017028692.1 6957 UTR 3 XP_016884181.1
XM_017028693.1 6957 UTR 3 XP_016884182.1
XM_017028694.1 6957 UTR 3 XP_016884183.1
XM_017028695.1 6957 UTR 3 XP_016884184.1
XM_017028696.1 6957 UTR 3 XP_016884185.1
XM_017028697.1 6957 UTR 3 XP_016884186.1
XM_017028698.1 6957 UTR 3 XP_016884187.1

View Full Product Details