Product Details
- SNP ID
-
rs1984388
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:31938035 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CATGGTCCAAAGACTTGTCTTTTGA[A/T]GCAGCCCTGTTGTATCCTCTTGAGT
- Phenotype
-
MIM: 113508
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C22orf24
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75883264] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C22orf24
- Gene Name
- chromosome 22 open reading frame 24
- Gene
- YWHAH
- Gene Name
- tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein eta
There are no transcripts associated with this gene.
View Full Product Details