Product Details

SNP ID
rs904411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:65170257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGATGGAGCAGTGGAGGACGAG[C/G]AAGGGGAGGGAGAGGACGGTGAGGA
Phenotype
MIM: 607844
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LEMD3 PubMed Links

Gene Details

Gene
LEMD3
Gene Name
LEM domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167614.1 687 Missense Mutation CAA,GAA Q221E NP_001161086.1
NM_014319.4 687 Missense Mutation CAA,GAA Q221E NP_055134.2

View Full Product Details