Product Details

SNP ID
rs10033
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55425095 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCATCCAAGAGGCATAGCAGCAG[C/T]AGAAGTCTTTAAAGGCTTGTACACC
Phenotype
MIM: 607124
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PLPP1 PubMed Links

Gene Details

Gene
PLPP1
Gene Name
phospholipid phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003711.3 3718 UTR 3 NP_003702.2
NM_176895.2 3718 UTR 3 NP_795714.1
XM_006714724.2 3718 UTR 3 XP_006714787.1
Gene
SKIV2L2
Gene Name
Ski2 like RNA helicase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015360.4 3718 UTR 3 NP_056175.3

View Full Product Details