Product Details
- SNP ID
-
rs14842
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:46243560 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAAAATGGCTATAAAATAACCTTA[A/T]TTTTAAAAAAAAATCTTGGGTCTTC
- Phenotype
-
MIM: 170998
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CDPF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77378083] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CDPF1
- Gene Name
- cysteine rich DPF motif domain containing 1
- Gene
- PPARA
- Gene Name
- peroxisome proliferator activated receptor alpha
View Full Product Details