Product Details
- SNP ID
-
rs1943382
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:108509608 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTGTGGTTGTGTCTGTGTCACAAT[C/T]TGAGTCCACTGGGTCATCCTCATAG
- Phenotype
-
MIM: 612878
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EXPH5
PubMed Links
Gene Details
- Gene
- EXPH5
- Gene Name
- exophilin 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001144763.1 |
6010 |
Missense Mutation |
AAT,GAT |
N1891D |
NP_001138235.1 |
NM_001144764.1 |
6010 |
Missense Mutation |
AAT,GAT |
N1811D |
NP_001138236.1 |
NM_001144765.1 |
6010 |
Missense Mutation |
AAT,GAT |
N1779D |
NP_001138237.1 |
NM_001308019.1 |
6010 |
Missense Mutation |
AAT,GAT |
N1960D |
NP_001294948.1 |
NM_015065.2 |
6010 |
Missense Mutation |
AAT,GAT |
N1967D |
NP_055880.2 |
XM_011542696.2 |
6010 |
Missense Mutation |
AAT,GAT |
N1891D |
XP_011540998.1 |
XM_011542698.2 |
6010 |
Missense Mutation |
AAT,GAT |
N1891D |
XP_011541000.1 |
XM_011542700.2 |
6010 |
Missense Mutation |
AAT,GAT |
N1891D |
XP_011541002.1 |
XM_017017397.1 |
6010 |
Missense Mutation |
AAT,GAT |
N1891D |
XP_016872886.1 |
XM_017017398.1 |
6010 |
Missense Mutation |
AAT,GAT |
N1890D |
XP_016872887.1 |
XM_017017399.1 |
6010 |
Missense Mutation |
AAT,GAT |
N1891D |
XP_016872888.1 |
XM_017017400.1 |
6010 |
Missense Mutation |
AAT,GAT |
N1891D |
XP_016872889.1 |
XM_017017401.1 |
6010 |
Missense Mutation |
AAT,GAT |
N1890D |
XP_016872890.1 |
XM_017017402.1 |
6010 |
Missense Mutation |
AAT,GAT |
N1811D |
XP_016872891.1 |
XM_017017403.1 |
6010 |
Missense Mutation |
AAT,GAT |
N1810D |
XP_016872892.1 |
XM_017017404.1 |
6010 |
Missense Mutation |
AAT,GAT |
N1779D |
XP_016872893.1 |
View Full Product Details