Product Details
- SNP ID
-
rs15040
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:106238787 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTAGAGACATCCATGAAAAGCCAC[A/C]TGTTATTCACAGGGGCTGCGCTTCA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CKAP4
PubMed Links
Gene Details
- Gene
- CKAP4
- Gene Name
- cytoskeleton associated protein 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006825.3 |
2130 |
UTR 3 |
|
|
NP_006816.2 |
View Full Product Details