Product Details
- SNP ID
-
rs1290005
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:94570381 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGTTTTCATTTCCTGAGTAAGCTT[C/T]GCAGTCCCTCACAGACCTGTGAATA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR5700
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114187034] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR5700
- Gene Name
- microRNA 5700
There are no transcripts associated with this gene.
- Gene
- MIR7844
- Gene Name
- microRNA 7844
There are no transcripts associated with this gene.
- Gene
- TMCC3
- Gene Name
- transmembrane and coiled-coil domain family 3
View Full Product Details