Product Details

SNP ID
rs1126712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:114324845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATCCTCAGGGACACCAAGACCTA[C/G]ATGCTTGCTTTTGACGTGAACGATG
Phenotype
MIM: 605729 MIM: 138600 MIM: 138610
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AKNA PubMed Links

Gene Details

Gene
AKNA
Gene Name
AT-hook transcription factor
There are no transcripts associated with this gene.

Gene
ORM1
Gene Name
orosomucoid 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000607.2 495 Nonsense Mutation TAC,TAG Y128* NP_000598.2
Gene
ORM2
Gene Name
orosomucoid 2
There are no transcripts associated with this gene.

View Full Product Details