Product Details

SNP ID
rs11151997
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:57147794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCACTCCCTCCAGAGCCAGAAG[G/T]CCAGGACCCTCCAGCTCCGTCTCAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BOD1L2 PubMed Links

Gene Details

Gene
BOD1L2
Gene Name
biorientation of chromosomes in cell division 1 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257964.1 733 Missense Mutation GGC,GTC G161V NP_001244893.1

View Full Product Details