Product Details

SNP ID
rs6781
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127935764 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACTCTCTTGGTCTTCAGCATCAC[A/G]TAGGAGATGAACAGTCCTGGGATAC
Phenotype
MIM: 603564 MIM: 610891 MIM: 612865
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPM2 PubMed Links

Gene Details

Gene
DPM2
Gene Name
dolichyl-phosphate mannosyltransferase polypeptide 2, regulatory subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003863.3 877 Silent Mutation NP_003854.1
Gene
FAM102A
Gene Name
family with sequence similarity 102 member A
There are no transcripts associated with this gene.

Gene
PIP5KL1
Gene Name
phosphatidylinositol-4-phosphate 5-kinase like 1
There are no transcripts associated with this gene.

View Full Product Details