Product Details

SNP ID
rs1869736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:9708084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGGGCGGATGCCGGAGGGCGC[C/T]GGGGGAGGCCGACCCGCGCGGGGCT
Phenotype
MIM: 605694
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB31 PubMed Links

Gene Details

Gene
RAB31
Gene Name
RAB31, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006868.3 114 Intron NP_006859.2
XM_017025528.1 114 UTR 5 XP_016881017.1
XM_017025529.1 114 UTR 5 XP_016881018.1

View Full Product Details