Product Details

SNP ID
rs2066577
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:50091308 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTATATATCATATTATTGACTGC[C/T]GGACTGCTTAACATGCAGCTTAATA
Phenotype
MIM: 605765 MIM: 605766
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DLEU1 PubMed Links

Gene Details

Gene
DLEU1
Gene Name
deleted in lymphocytic leukemia 1 (non-protein coding)
There are no transcripts associated with this gene.

Gene
DLEU2
Gene Name
deleted in lymphocytic leukemia 2 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details